accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOONOOOOHH3COCH3HOCH3CH3OHOCH3H3CH3CH3COH3C(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 53123-88-9

IUPAC Name: 1,18-dihydroxy-12-[1-(4-hydroxy-3-methoxycyclohexyl)propan-2-yl]-19,30-dimethoxy-15,17,21,23,29,35-hexamethyl-11,36-dioxa-4-azatricyclo[30.3.1.0⁴,⁹]hexatriaconta-16,24,26,28-tetraene-2,3,10,14,20-pentone
Synonyms: sirolimus
Molecular Weight (g/mol): 914.19
Molecular Formula: C51H79NO13
InChi Key: QFJCIRLUMZQUOT-UHFFFAOYNA-N
SMILES: COC1CC(CC(C)C2CC(=O)C(C)C=C(C)C(O)C(OC)C(=O)C(C)CC(C)C=CC=CC=C(C)C(CC3CCC(C)C(O)(O3)C(=O)C(=O)N3CCCCC3C(=O)O2)OC)CCC1O
Color 
  • (7)
  • (1)
Molecular Weight (g/mol) 
  • (7)
  • (1)
Physical Form 
  • (1)
  • (6)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Type 
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (7)
  • (1)
  • (7)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (8)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)