Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAPGEF5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567436
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567436 Supplier Invitrogen™ Supplier No. PA567436
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DFLLTYTVFMT TDDLCQALLR HYSAKKYQGK EENSDVPRRK RKVLHLVSQW IALYKDWLPE DEHSKMFLKT IYRNVLD Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.

Members of the RAS subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as Repac, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation. Repac, also known as Rap guanine nucleotide exchange factor 5 (RAPGEF5), is a 580 amino acid protein that is expressed in brain and testis, with weaker expression in heart, placenta, lung, pancreas and small intestine. Existing as two alternatively spliced isoforms, the Repac gene is conserved in cow, mouse and rat. The gene that encodes Repac maps to human chromosome 7p15.3, with high expression of this region linking to positive response to erlotinib therapy for those with non-small-cell lung cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAPGEF5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAPGEF5
Gene Accession No. Q92565
Gene Alias 4932413M22; C86120; D030051B22Rik; GFR; guanine nucleotide exchange factor; guanine nucleotide exchange factor for Rap1; KIAA0277; mKIAA0277; mmr-gef; Mras regulated guanine nucleotide exchange factor; M-Ras-regulated GEF; M-Ras-regulated Rap GEF; MRGEF; MR-GEF; Rap guanine nucleotide exchange factor (GEF) 5; Rap guanine nucleotide exchange factor 5; RAPGEF5; Related to Epac; REPAC; RGD1562891; rMR-GEF; tcag7.1034
Gene Symbols RAPGEF5
Host Species Rabbit
Immunogen Recombinant Human RAPGEF5. Recombinant protein control fragment (Product #RP-108097).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9771
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.