Learn More
3M™ Petrifilm™ Rapid Aerobic Count Plate, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S30758 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
50 Pack
500 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30758 500 Pack
S30757 50 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S30758 Supplier 3M™ Petrifilm™ Supplier No. 6479
Add to Cart
Add to Cart
Technology facilitates colony enumeration in 24 hours for most food matrices/environmental samples and resists distortion caused by spreader colonies
  • Plates are sample-ready, eliminating time-consuming, cumbersome step of preparing media/agar dishes
  • Provides accurate, efficient, consistent results for food processors and contract labs
  • Can be used with 3M™ Petrifilm™ Plate Reader for consistent, fast, automated reading and recording of results
  • This plate has been awarded Official Methods of Analysis (OMA) #2015.13.for Selected Food NF VALIDATION 3M 01/17-11/16 for Dairy products by AFNOR Certification
Specifications
Product Type Count Plate
Test Type Environmental Test
For Use With (Application) Food matrices and environmental samples, Facilitating colony enumeration
For Use With (Equipment) 3M™ Petrifilm™ plate reader
Quantity 500 Pack