Learn More
3M™ Petrifilm™ Rapid Coliform Count Plate, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. p-7232122 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
50 Pack
500 Pack
Product Type:
Count Plate
Test Type:
Environmental Test
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Test Type
S30723 50 Pack Count Plate Environmental Test
S30690 500 Pack Count Plate Environmental Test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S30723 Supplier 3M™ Petrifilm™ Supplier No. 6402
Only null left
Add to Cart
Add to Cart
Designed to provide fast, accurate results in detecting coliform contamination
  • Proven cost effective tool for detection of coliform
  • Provides accurate, efficient, consistent results for food processors and contract labs
  • Results in 4 to 24 hours
  • This plate has been awarded Official Methods of Analysis (OMA) #2000.15 for Food and NF VALIDATION certificates: 3M-01/5-03/97 A for all human food products , 3M-01/5-03/97 B for All human food products and 3M-01/5-03/97 C for All human food products (except process pork products) by AFNOR Certification
Specifications
Detects Coliform
Product Type Count Plate
Test Type Environmental Test
For Use With (Application) Food processors and contract labs
Quantity 50 Pack