Learn More
3M™ Petrifilm™ Rapid E.coli/Coliform Count Plate
SCI_ED
Catalog No. p-7232134 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
50 Pack
500 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30759 50 Pack
S30760 500 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S30759 Supplier 3M™ Petrifilm™ Supplier No. 6436
Only null left
Add to Cart
Add to Cart
Provides cost-effective, convenient and reliable method for testing equipment, raw materials, food products and manufacturing environmental samples
  • E. coli and coliform results in 18 to 24 hours
  • Provides accurate, efficient, consistent results for food processors and contract labs
  • Plates are sample-ready, eliminating time-consuming, cumbersome step of preparing media/agar dishes
  • Can be used with 3M™ Petrifilm™ Plate Reader for consistent, fast, automated reading and recording of results
  • This plate has been awarded Official Methods of Analysis (OMA) #2018.13 for select food and MicroVal certificate no. 2017LR76 for a broad range of foods and select environmental surfaces by Lloyd's Register Nederland B.V. following ISO 16140-2. Food Categories tested: meat products, dairy and egg products, fish and seafood products, vegetable products, ready to eat and ready to reheat, animal feed and samples from the industrial production environment
Specifications
Detects E. coli and coliform
Product Type Count Plate
Test Type Environmental Test
For Use With (Application) Testing equipment, raw materials, food products and manufacturing environmental samples, Food processors and contract labs
For Use With (Equipment) 3M™ Petrifilm™ plate reader
Quantity 50 Pack