Learn More
XpressBio Rapid-Lenti HIV-1 p24 ELISA Kit (Single Wash) 96 wells
SDP
Supplier:  XpressBio XB4000
Encompass_Preferred
The Rapid-Lenti HIV-1 p24 ELISA Kit (Single Wash) can be used to monitor HIV-1 in cell culture or to determine the viral titer of lentiviral samples and can be completed in as little at 90 minutes.
Catalog No. NC2333877
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.