Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Rapid-RT MasterMix (5X)

Catalog No. MP116187400 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116187400 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116187400 Supplier MP Biomedicals™ Supplier No. 116187400
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapid-RT MasterMix (5X) is a unique reverse transcriptase mastermix buffer that generates consistent first-strand cDNA from as little as 1 pg of starting total RNA.

The all-in-one Rapid-RT-MasterMix (5X) is a ready to use formulation for simple and reliable first-strand cDNA synthesis with the ability to remove gDNA, without affecting reverse transcription. The resulting cDNA is compatible with any number of downstream applications.

  • Easy one-step setup reduces pipetting and sample handling
  • High cDNA yields from difficult samples
  • Removes contaminating gDNA with temperature-sensitive DNase
  • Can reverse transcribe RNA of low abundance or degraded templates
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Product Type Reverse Transcriptase
Reverse Transcriptase Yes
For Use With (Application) Standard PCR
Hot Start Yes
No. of Reactions 100 Reactions
Reaction Format Mastermix
Includes 5X RT MasterMix (RapidScript Hot-Start Reverse Transcriptase, RNaseOFF Ribonuclease Inhibitor, temperature-sensitive DNase, dNTPs, optimal ratio of oligo(dT) and random primers), 400 μL; Nuclease-free Water, 1.0 mL
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.