Learn More
LABSYSTEMS DIAGNOSTICS OY Rapid test for detection of IgM antibodies
Supplier:  LABSYSTEMS DIAGNOSTICS OY 3034000
Encompass_Preferred
Biocard™ Chlamydia pneumoniae IgM test is a rapid immunochromatographic test that detects anti-Chlamydia pneumoniae IgM antibodies from a blood sample. If the sample contains these antibodies, they will bind with the gold labelled antibodies and with the stationary reagent in the test line. The test also contains an integrated control system. A red control line indicates the proper function of the test. The test requires only one drop of fingertip blood (10 μl) or serum (5 μl) and it can be performed and evaluated in 5 to 10 minutes.
Catalog No. NC2865486
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.