Learn More
LABSYSTEMS DIAGNOSTICS OY Rapid test for detection of IgM antibodies
Supplier:  LABSYSTEMS DIAGNOSTICS OY 3035000
Encompass_Preferred
Biocard™ Mycoplasma pneumoniae IgM test is a rapid immunochromatographic test that detects anti-M. pneumoniae IgM antibodies. If the sample contains these antibodies, they will bind to labelled antigen and with the stationary reagent in the test line. The test also contains an internal control system. Appearance of a red control line indicates proper functioning of the test. The test requires one drop (10μl) of blood or 5 μl of serum and it can be performed and evaluated in 10 minutes. A positive test result with this test is an aid for diagnosis of acute M.pneumoniae infection.
Catalog No. NC2865502
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.