Learn More
3M™ Petrifilm™ Rapid Yeast and Mold Count Plate, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S30718 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
500 Pack
Catalog No. Quantity
S30718 500 Pack
1 options
Catalog No. S30718 Supplier 3M™ Petrifilm™ Supplier No. 6477
Add to Cart
Add to Cart
New indicator technology simplifies and accelerates detection, makes colonies easier to interpret, results in 48 hours
  • Utilizes convenience and efficiency of 3M™ Petrifilm™ plate platform
  • Proven cost effective tool for detecting yeast and mold
  • Proven as reliable as DRBC and DG-18 spread plate methods, as well as acidified PDA and YGC agar pour plate methods
  • Can be used with both low and high water activity foods
  • Eliminates need for labor intensive, multiple plate spread plate technique
  • Versatility for air, swab or surface contact environmental sampling
Specifications
Detects Yeast and mold
Product Type Count Plate
Test Type Environmental Test
Quantity 500 Pack