Learn More
Solar Biologicals Inc RapidChek Listeria Strips, 450 Strips Per Pack, Results In 10 Minutes
Supplier:  Solar Biologicals Inc 10001372
Encompass
The RAPIDCHEK LISTERIA STR 450/PK offers a quick and reliable solution for detecting *Listeria* in food and environmental samples. With results in just 10 minutes and AOAC certification, it enhances food safety protocols by significantly reducing turnaround time compared to traditional methods. Compatible with various enrichment media, this product distinguishes itself with its double antibody sandwich format, making it suitable for diverse testing environments.
Catalog No. NC3591185