Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers RAPIDLab™ 1200 Series Ready Sensor™ Biosensors/Electrodes

Catalog No. 23044828 Shop All Siemens Healthineers Products
Change view
Click to view available options
Sensor Types:
Calcium Biosensor
Chloride Biosensor
Lactate Biosensor
Potassium Biosensor
Sodium Biosensor
pCO2 Biosensor
pH Biosensor
pO2 Biosensor
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sensor Types
23-044-828 Lactate Biosensor
23-044-841 Calcium Biosensor
23-044-866 Chloride Biosensor
23-044-849 pCO2 Biosensor
23-044-863 pH Biosensor
23-044-848 pO2 Biosensor
23-044-865 Potassium Biosensor
23-044-864 Sodium Biosensor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 23-044-828 Supplier Siemens Healthineers Supplier No. 10311032
Add to Cart
Add to Cart
This item is not returnable. View return policy

An in vitro diagnostic device for use on the RAPIDLab™ 1200 series instruments.

TRUSTED_SUSTAINABILITY

Specifications

Sensor Types Lactate Biosensor
For Use With (Equipment) RAPIDLab™ 1200 Series Instruments
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.