Learn More
Siemens Healthineers RAPIDLyte Multicap™-S Plastic Blood Collection Capillary Tubes
Shop All Siemens Healthineers ProductsDescription
Multicap™-S capillary tubes are available in a variety of sizes and configurations to meet your clinical needs across a range of critical care settings.
- Choose from a variety of capillaries in multiple configurations to meet your arterial blood sampling needs
- Maintain the highest level of sample integrity with no dilution errors, minimized electrolyte binding and fast, easy air bubble removal
- Maximize your instrument uptime with our advanced, dry-sprayed, balanced and reduced heparin formulations to minimize the potential for clots
Specifications
Specifications
| Product Type | Blood Collection Capillary Tubes |
| Material | Plastic |
| Volume (Metric) | 140 μL |
| For Use With (Application) | pH/Blood Gas/Na+/K+/Ca++/Cl-/Glu/Lac/tHb Testing |
| Quantity | 500 Tubes |