Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Rappaport-Vassiliadis Enrichment Broth (Dehydrated)

Catalog No. OXCM0669B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0669B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0669B Supplier Thermo Scientific™ Supplier No. CM0669B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively enrich Salmonella spp. from food and environmental specimens with this selective medium.

Selectively enrich Salmonella spp. from food and environmental specimens with Thermo Scientific™ Oxoid™ Rappaport-Vassiliadis Enrichment Broth (Dehydrated). Rappaport-Vassiliadis Enrichment Broth selects for Salmonella species by exploiting their characteristics when compared with other Enterobacteriaceae, such as the ability to survive at relatively high osmotic pressure, multiply at relatively low pH values, be relatively more resistant to malachite green, and have relatively less demanding nutritional requirements1.

Salmonella are a group of closely related rod-shaped, Gram-negative bacteria. Salmonella typically live in animal and human intestines and are shed through feces. Humans become infected most frequently through contaminated water or food. The most common symptoms of Salmonella infection are diarrhea, fever, abdominal cramps, and vomiting.

Use Rappaport-Vassiliadis Enrichment Broth for selective enrichment of Salmonella species food and environmental samples.

  • Selective: malachite green, low pH, and high magnesium chloride concentration inhibit many Gram-positive bacteria, while selectively enriching for salmonellae.
  • Superior performance: superior performance to Selenite Enrichment Broth and Tetrathionate Broth for the enrichment of salmonellae with the exception of Salmonella Typhi1.

Rappaport-Vassiliadis (RV) Enrichment Broth is similar to that described by Vassiliadis et al.3 except that the peptone used is soya peptone, which has been reported to enhance the growth of salmonellae1,2.

Not all products are available for sale in all territories. Please inquire.


Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Rappaport-Vassiliadis Enrichment Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 16.7L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.