Learn More
Description
MA1-810A detects retinoic acid receptor (RAR) alpha from human, rat and mouse tissues and overexpressed cells. This antibody does not detect any of the RAR beta or gamma forms. MA1-810A has been successfully used in Western blot and ICC/IF procedures. By Western blot, this antibody detects a single ~45 kDa protein representing RAR alpha in transfected COS cells. Due to the extremely low levels of RAR expressed in native tissues, it is recommended that sensitive detection systems, such as enhanced chemiluminescence, be used. The apparent molecular weight of RAR alpha from transfected COS cells is ~45 kDa. In rat tissues, such as skin and testis, doublets in the ~50 kDa area are seen. RAR alpha has been identified with this antibody in human dermal and neonatal fibroblast cell lines, as well as mouse F9 and human SH-SY5Y cells; however, RAR alpha from human leukocytes and HUT102 cells were not identified by MA1-810 suggesting that there may be structural variation of RAR alpha between some cell types. The MA1-810A immunogen is a synthetic peptide corresponding to residues C(444) S P S L S P S S H R S S P A T Q S P(462) of mouse RAR alpha. This peptide (Cat. # PEP-004) is available for use in neutralization and control experiments. Reconstitute with PBS.
Specifications
Specifications
| Antigen | RARA |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | Ralpha10 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.05% sodium azide |
| Gene | RARA |
| Gene Accession No. | P10276, P11416 |
| Gene Alias | Nr1b1; nuclear receptor subfamily 1 group B member 1; nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form; RAR; RAR Alpha; RAR alpha 1; RARA; RAR-alpha; RARalpha1; retinoic acid; retinoic acid nuclear receptor alpha variant 1; retinoic acid nuclear receptor alpha variant 2; retinoic acid receptor; retinoic acid receptor alpha; retinoic acid receptor, alpha; retinoic acid receptor, alpha polypeptide; RRA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
