Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RARB Monoclonal Antibody (R beta 28)
GREENER_CHOICE

Catalog No. MA1811 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1811 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1811 Supplier Invitrogen™ Supplier No. MA1811
  • $563.93
    $423.65 / Each of 1

    Save $140.28

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-811 detects retinoic acid receptor (RAR) beta from human and mouse tissues. This antibody cross-reacts slightly with RAR alpha when expressed in COS cells, though the level of signal on Western blots is very low relative to that seen with RAR beta. It does not detect RAR alpha or gamma from cell lines or tissue extract. MA1-811 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~52 kDa protein in rat tissue extract representing RAR beta. Also, by Western blot MA1-811 detects an unidentified ~120 kDa protein which can be competed away with the immunizing peptide. This protein is most notable in human SH-SY5Y and COS cells. Due to the extremely low levels of RAR expressed in native tissues, it is recommended that sensitive detection systems, such as enhanced chemiluminescence, be used. The MA1-811 immunogen is a synthetic peptide corresponding to the C-terminal sequence of mouse RAR beta. Reconstitute with PBS. MA1-811 can be used with blocking peptide PEP-005.

Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RARB
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone R beta 28
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene RARB
Gene Accession No. P10826, P22605
Gene Alias A830025K23; HAP; HBV-activated protein; hepatitis B virus activated protein; MCOPS12; NR1B2; nuclear receptor subfamily 1 group B member 2; RAR beta; Rarb; RAR-b; RAR-beta; RARbeta1; RAR-Beta2; RAR-Beta4; RAR-epsilon; retinoic acid; retinoic acid receptor; retinoic acid receptor beta; retinoic acid receptor, beta; retinoic acid receptor, beta polypeptide; RRB2
Gene Symbols RARB
Host Species Mouse
Immunogen Synthetic peptide corresponding to the C-terminal sequence of the mouse RAR beta.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218772, 24706, 5915
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.