Learn More
QuidelOrtho Rare Antisera
SDP
Catalog No. 23287319
Change view
Click to view available options
Quantity:
3 mL
5 mL
Type:
Antisera Anti-Fya (Duffy)
Antisera Anti-Jka BioClone
Antisera Anti-Jkb BioClone
Antisera Anti-Kell
Antisera Anti-M (Rabbit)
Antisera Anti-S
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-287-319 5 mL Antisera Anti-M (Rabbit)
23-287-461 3 mL Antisera Anti-Jka BioClone
23-287-462 3 mL Antisera Anti-S
OK715130 3 mL Antisera Anti-Fya (Duffy)
23-287-322 5 mL Antisera Anti-Kell
23-287-323 3 mL Antisera Anti-Jkb BioClone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 23-287-319 Supplier QuidelOrtho Supplier No. 715850
Add to Cart
Add to Cart
This item is not returnable. View return policy
  • Antisera are used as qualitative tests for recognition of antigens on human red blood cells
  • Each antisera is used to indicate the presence of specific antigens, which when present will cause clumping of normal human red blood cells
Specifications
Type Antisera Anti-M (Rabbit)
For Use With (Application) For recognition of Antigens on Human red Blood cells
Quantity 5 mL