Description
Includes
- Ras Assay Reagent (Raf-1 RBD, agarose)
- Anti-Ras, clone RAS10
- 100X GTPγS, 10mM
- 100X GDP, 100mM
- Mg2+ Lysis/Wash Buffer, 5X
Specifications
Specifications
| For Use With (Application) | Enzyme Assay |
| Quantity | 30 Assays/Kit |
| Type | Activation Assay Kit |
| Cell Line | HeLa |