Learn More
Strem, An Ascensus Company CAS . 25mg. (R)-()-7-[4(S)-(Benzyl)oxazol-2-yl]-7-di(35-di-t-butylphenyl)phosphino-2233-tetrahydro-11-spirobiindane min. 97% (RaS)-DTB-Bn-SIPHOX. MFCD17018765
SDP
Supplier:  Strem, An Ascensus Company 15519025MG
Encompass_Preferred
25mg. (R)-()-7-[4(S)-(Benzyl)oxazol-2-yl]-7-di(35-di-t-butylphenyl)phosphino-2233-tetrahydro-11-spirobiindane min. 97% (RaS)-DTB-Bn-SIPHOX. MFCD17018765. Molecular Weight 788.09. Molecular Formula C55H66NOP. Color/form white solid Strem 15-5190. http//www.strem.com/catalog/v/15-5190/
Catalog No. 50-211-0769
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.