Learn More
Active Motif Ras GTPase Chemi ELISA Kit
SDP
Supplier:  Active Motif 52097
Encompass_Preferred
Active Motif's Ras GTPase Chemi ELISA Kit is the first ELISA-based kit designed to detect and quantify activated Ras GTPase. Because of its ELISA format, it is much more sensitive than pull-down/Western methods. This enables the detection of low levels of protein, while also using less sample. In addition, ELISAs are more quantitative than Westerns, and they eliminate the need to run and develop gels.
Catalog No. NC0280362
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.