Learn More
TARGETMOL CHEMICALS INC Spautin-1 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T193725MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Spautin-1 is an autophagy inhibitor that suppresses the deubiquitination activity of ubiquitin-specific peptidase USP10 and USP13. Spautin-1 inhibits UPR and exhibits preferential cytotoxicity in glucose-starved cancer cells. Purity 98.99%
Catalog No. 50-287-8302
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.