Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASA1 Monoclonal Antibody (B4F8)
GREENER_CHOICE

Catalog No. MA4001 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA4001 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA4001 Supplier Invitrogen™ Supplier No. MA4001
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA4-001 detects GTPase activating protein (GAP) from human, mouse, rat, bovine and non-human primate tissues. MA4-001 has been successfully used in Western blot, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody recognizes a single 120 kDa protein representing ras GAP from RS-2 cell lysate. Immunofluorescence staining of GAP in mouse fibroblast cells with MA4-001 results in diffuse cytoplasmic staining. Following the addition of PDGF, immunofluorescence staining shows that some GAP rapidly translocates to the plasma membrane. MA4-001 immunoprecipitates GAP that is complexed with the GAP related p190. The MA4-001 antigen is full length recombinant human GAP. Epitope mapping studies suggest that this antibody binds a portion of GAP that contains the src homology regions SH2 & SH3. Reconstitute with PBS.

TACC1 is located on 8p11 chromosomal region that is amplified in approximately 15% of all breast tumor samples. The short arm of chromosome 8 also contains FGFR1 whose expression is enhanced in most breast cancer tumors. TACC family members, TACC1, TACC2, and TACC3, map very closely to the corresponding FGFR1, FGFR2, FGFR3 genes on chromosomes 4,8, and 10. Subsequently, since they are phylogenetically related, it is proposed that TACC and FGFR have similar roles in cell growth and differentiation. Also, TACC1 contains a conserved C-terminal region as in the Drosophila homolog, D-TACC. It has been shown that D-TACC is necessary for normal spindle function, and the mammalian TACC proteins appears to interact with centrosomes and microtubules in a similar manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASA1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone B4F8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene RASA1
Gene Accession No. P09851, P20936, P50904
Gene Alias CMAVM; CM-AVM; Gap; GAPX; GTPase-activating protein; p120; p120 RAS GTPase activating protein; p120GAP; p120RASGAP; p120-rasGAP; PKWS; Ras GTPase-activating protein 1; ras GTPase-activating protein 1-like protein; Ras p21 protein activator; RAS p21 protein activator (GTPase activating protein RAS p21); RAS p21 protein activator (GTPase activating protein) 1; RAS p21 protein activator 1; Rasa; RASA1; RASGAP; triphosphatase-activating protein; unnamed protein product
Gene Symbols RASA1
Host Species Mouse
Immunogen Full length recombinant human GAP.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218397, 25676, 282032, 5921
Target Species Bovine, Human, Mouse, Monkey, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.