Learn More
AdipoGen RasGRP2 pSer587 pAb IG1209
SDP
Supplier:  AdipoGen AG25T0119C020
Encompass_Preferred
Polyclonal Antibody. Recognizes human and mouse RasGRP2 phosphorylated at pSer587 human isoform 1, pSer586 mouse or pSer649 human isoform 2. Source Rabbit. Applications WB. Liquid. In PBS containing 2mg/ml BSA and 0.05% sodium azide. RasGRP2 functions as a calcium- and DAG-regulated nucleotide exchange factor specifically activating small GTPases, including Rap1/RAS3, through the exchange of bound GDP for GTP. It may also activate other GTPases such as RRAS, RRAS2, NRAS, KRAS but not HRAS. The protein is expressed in platelets, neutrophils and T lymphocytes. RasGRP2 functions in aggregation of platelets and adhesion of T lymphocytes and neutrophils probably through inside-out integrin activation. S587 was identified as the major PKA phosphorylation. Phosphorylation of S587 correlated with the inhibitory effect of PKA on Rap1b activation in platelets.
Catalog No. 50-256-3155
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.