Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASGRP3 Mouse anti-Human, Mouse, Clone: 1A10G4, Proteintech
SDP

Catalog No. 501737068 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7068 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7068 Supplier Proteintech Group Inc Supplier No. 666501IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Members of the RAS.

Specifications

Antigen RASGRP3
Applications Western Blot
Classification Monoclonal
Clone 1A10G4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RASGRP3
Gene Accession No. Q8IV61
Gene Alias GRP3, KIAA0846, Ras guanyl releasing protein 3, RASGRP3
Gene Symbols Rasgrp3
Host Species Mouse
Immunogen RASGRP3 Fusion Protein Ag3810
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240168, 25780
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.