Learn More
Apexbio Technology LLC Anti-IL-7Ra/CD127 Antibody (Lusvertikimab) 5mg
Supplier: Apexbio Technology LLC F12165
Lusvertikimab (Anti-IL-7Ra CD127 Antibody CAS 2375835-91-7) is a humanized monoclonal antibody targeting the interleukin-7 receptor alpha chain (IL-7Ra/CD127) It is designed to inhibit IL-7 receptor signaling by preventing heterodimerization of IL-7Ra with the common gamma chain ( c) thereby blocking downstream signaling pathways Lusvertikimab exerts its biological activity primarily by binding IL-7Ra without inducing receptor internalization In studies of B-cell precursor acute lymphoblastic leukemia (BCP-ALL) Lusvertikimab demonstrates inhibitory effects on IL-7-mediated signaling Based on these pharmacological properties Lusvertikimab holds research potential in the study of leukemia biology and the investigation of IL-7 receptor signaling in hematological malignancies
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.