Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASSF1A Monoclonal Antibody (eB114-10H1), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5013559
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-135-59 100 μg
50-135-58 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-135-59 Supplier Invitrogen™ Supplier No. 14688882
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: The eB114 antibody reacts with the RASSF1A isoform of the tumor suppressor gene RASSF1. RASSF1A contains a Ras Association Domain (RA) and binds Ras in a GTP-dependent manner to effect apoptosis. The RASSF1A isoform undergoes epigenetic inactivation in a variety of human primary tumors. This has been associated with hypermethylation of a CpG island(s) in its promoter region. This antibody was generated against a synthetic peptide corresponding to the splice junction region of RASSF1A. Applications Reported: The eB114 antibody has been reported for use in immunoprecipitation, and immunoblotting (WB). Applications Tested: The eB114-10H1 has been tested by Western blotting and can be used at approximately 1-10 μg/mL. RASS1A is detected as a 45 kDa band in Western blotting of a HeLa cell extract. It is recommended that the reagent be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

This gene encodes a protein similar to the RAS effector proteins. Loss or altered expression of this gene has been associated with the pathogenesis of a variety of cancers, which suggests the tumor suppressor function of this gene. The inactivation of this gene was found to be correlated with the hypermethylation of its CpG-island promoter region. The encoded protein was found to interact with DNA repair protein XPA. The protein was also shown to inhibit the accumulation of cyclin D1, and thus induce cell cycle arrest. Seven alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF1A
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone eB114-10H1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene RASSF1
Gene Accession No. Q9NS23
Gene Alias 123F protein; 123F2; AA536941; AU044980; cardiac-specific ras association domain family 1 protein; NORE2A; pancreas-specific ras association domain family 1 protein; protein 123F2; Ras association (RalGDS/AF-6) domain family 1; Ras association (RalGDS/AF-6) domain family member 1; Ras association domain family 1; Ras association domain family member 1; ras association domain-containing protein 1; Rassf1; Rassf1A; RASSF1A tumor suppressor; Rassf1B; Rassf1C; RDA32; REH3P21; tumor suppressor protein RDA32; wu:fc17f02; WUGSC:H_LUCA12.5; zgc:112224; zgc:92505; zgc:92505 protein
Gene Symbols RASSF1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11186
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.