Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASSF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592352
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592352 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592352 Supplier Invitrogen™ Supplier No. PA592352
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTMMAHQYPS WIFINEKTFI TREQLNSLLK TYNIFYENQK NLHILYGETE DGKLIVEGML DIFWGVKRPI QLKIQDEKPF SSFTSMKSSD VFSSKGMTRW GEFDDLYRIS ELDRTQIPMS EKRNSQEDYL SYHSNTLKPH AKDEPDSPVL YRTMSEAALV RKRMKPLMMD RKERQKNRAS INGHFYNHEI ENSPQDFALH IIFATGEQRR LKKTDIPLLQ RLLQGPSEKN ARIFLMDKDA EEISSDVAQY INFHFSLLES ILQRLNEEEK REIQRIVTKF NKEKAIILKC LQNKLVIKTE TTV; Positive Samples: Mouse intestine, Rat kidney.

RASSF6 is a member of the RAS association domain family of RAS effectors/tumor suppressors that mediate some of the growth inhibitory properties of RAS oncogenes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rassf6
Gene Accession No. Q4QR82, Q80UQ2
Gene Alias 1600016B17Rik; DKFZp686K23225; putative RAS binding protein; Ras association (RalGDS/AF-6) domain family 6; Ras association (RalGDS/AF-6) domain family member 6; Ras association domain family member 6; ras association domain-containing protein 6; RASSF6
Gene Symbols Rassf6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-303 of human RASSF6 (NP_0012573201).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 305251, 73246
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.