Learn More
Biomatik Corporation Rat 11-Beta-Hydroxysteroid Dehydrogenase Type 2 (HSD11b2) ELISA Kit
Supplier:  Biomatik Corporation EKU02005
Encompass_Preferred
Rat 11-Beta-Hydroxysteroid Dehydrogenase Type 2 (HSD11b2) ELISA Kit for detecting HSD11b2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50233. Gene ID: 25117. Target Synonyms: AME; AME1; HSD11K; HSD2; SDR9C3; Short Chain Dehydrogenase/Reductase Family 9C,Member 3; NAD-dependent 11-beta-hydroxysteroid dehydrogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7562
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.