Learn More
Instech Rat 2ch VAB tether kit: VABR2T/25, red PinPort, 375/22P, 24in CoEx, LS22
SDP
Supplier:  Instech KVABR2T
Encompass_Preferred
Two channel Vascular Access Button tether kit for rats. 12in spring tether, red PinPort on intermittent line, 375/22PS swivel on continuous line, 24in COEX tubing and luer stub. Sterile. Connects to VABD95BS rat buttons.
Catalog No. NC1607597
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.