Learn More
Crystal Chem Inc Rat Albumin ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80662
Encompass_Preferred
The Rat Albumin ELISA is used to quantify levels of rat albumin in serum, plasma, and urine using only a 2 µL sample volume. Highlights: Kit uses sample size of only 2 µL. High sensitivity of 1.62 ng/mL. Dynamic range of 6.25 - 400 ng/mL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7791
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.