Learn More
Alpco Ltd Rat Alpha 2 Macroglobulin ELISA
SDP
Supplier:  Alpco Ltd 41A2MRTE01
Encompass
The Alpha 2-Macroglobulin ELISA is a highly sensitive two-site enzyme-linked immunoassay (ELISA) for the determination of Alpha 2-Macroglobulin in rat serum and plasma. For research use only. Not for use in diagnostic procedures.
Catalog No. NC2885131
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.