Learn More
Biomatik Corporation Rat Angiotensin III (AngIII) ELISA Kit
Supplier:  Biomatik Corporation EKU10635
Encompass_Preferred
Rat Angiotensin III (AngIII) ELISA Kit for detecting AngIII in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.67pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Ang-III; Angiotensin-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8321
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.