Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD115 Rat anti-Human, BB515, Clone: 9-4D2-1E4, BD

Catalog No. BDB565346 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565346 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565346 Supplier BD Biosciences Supplier No. 565346
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 9-4D2-1E4 monoclonal antibody specifically binds to CD115 which is also known as Colony stimulating factor 1 receptor (CSF-1R) or Macrophage colony-stimulating factor 1 receptor (M-CSFR). This type I transmembrane glycoprotein is a receptor tyrosine kinase (RTK) that belongs to the Ig superfamily. It is expressed on a variety of cells including those committed to the mononuclear phagocyte lineage, such as, monocytes, macrophages, and osteoclasts. CSF-1 binds to and signals through CSF-1R homodimers which undergo tyrosine autophosphorylation and transduce downstream signaling pathways resulting in cytoskeletal reorganization and gene expression. CSF-1R activation stimulates the proliferation, differentiation, and survival of cells within the mononuclear phagocyte system. Acting through CD115, CSF-1 induces macrophage spreading and motility, and in combination with RANKL, CSF-1 drives the differentiation of mononuclear phagocytes to become osteoclasts. Interleukin-34 (IL-34) is another ligand for CD115 that can induce similar, as well as, some different biological responses by CD115-positive target cells.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD115
Applications Flow Cytometry
Classification Monoclonal
Clone 9-4D2-1E4
Conjugate Brilliant Blue 515 (BB515)
Description CSF-1R; CSF1R; C-FMS; FMS; FIM2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Human CD115 Transfected Cell Line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.