Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Itgav, Rat, Clone: RMV-7, Abnova™

Catalog No. 89109159 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-159 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-159 Supplier Abnova Corporation Supplier No. MAB10946

Rat monoclonal antibody raised against Itgav.

Specifications

Antigen Itgav
Applications Flow Cytometry, Functional Assay, Immunoprecipitation
Classification Monoclonal
Clone RMV-7
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Itgav.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Gene Itgav
Gene Alias 1110004F14Rik/2610028E01Rik/CD51/D430040G12Rik
Gene Symbols Itgav
Host Species Rat
Immunogen Mouse Itgav.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16410
Test Specificity The monoclonal antibody reacts with the mouse Itgav, the integrin alpha v chain. This approximately 120 kDa surface molecule non-covalently associates with the beta subunits of the integrin family including beta 3 (CD61), beta 1 (CD29), beta 5 and beta 6 to form receptors for extracellular matrix components.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.