Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49d Rat anti-Mouse, Brilliant Violet 711, Clone: R1-2, BD Optibuild™

Catalog No. BDB740661 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740661 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740661 Supplier BD Biosciences Supplier No. 740661
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The R1-2 monoclonal antibody specifically binds to α 4 chain (CD49d), which is expressed as a heterdimer with either of two β , β 1 or β 7 (also known as β p). The α 4β α 4β 7 also preferentially binds to the mucosal vascular addressin, MAdCAM-1. The R1-2 antibody blocks some α 4 integrin-mediated binding functions. In combination with mAb 9C10 (MFR4.B) (Cat. No. 553313), binding of VLA-4 expressing cells to VCAM-1 can be almost completely inhibited.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49d
Applications Flow Cytometry
Classification Monoclonal
Clone R1-2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Description Integrin α4; Itga4; Integrin alpha-4; LPAM alpha; VLA-4 alpha; VLA-4a
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen AKR/Cum mouse spontaneous T lymphoma line TK1
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.