Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49e Rat anti-Mouse, Brilliant Violet 421, Clone: 5H10-27 (MFR5), BD Optibuild™

Catalog No. BDB740017 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740017 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740017 Supplier BD Biosciences Supplier No. 740017
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

The 5H10-27 (MFR5) monoclonal antibody specifically binds to the α5 chain of the integrin α5β1 fibronectin receptor (CD49e/CD29, VLA-5) on thymocytes, activated T lymphocytes, precursor and activated B cells, mast cells, platelets, epithelial and endothelial cells, fibroblasts and a variety of mouse cell lines. Soluble 5H10-27 (MFR5) antibody has been reported to inhibit VLA-5-mediated functions in vitro. In addition, immobilized 5H10-27 (MFR5) antibody reportedly costimulates the proliferative response of CD8+ T cells to plate-bound anti-CD3e antibody.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49e
Applications Flow Cytometry
Classification Monoclonal
Clone 5H10-27 (MFR5)
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Itga5; ITA5; Integrin alpha-5; Fibronectin receptor alpha; Fnra; VLA5 alpha
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Mast Cell Line MC/9
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.