Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD51 Rat anti-Mouse, Brilliant Violet 421, Clone: RMV-7, BD Optibuild™

Catalog No. BDB740062 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740062 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740062 Supplier BD Biosciences Supplier No. BDB740062
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The RMV-7 monoclonal antibody specifically binds to CD51, the 140 kDa integrin α V chain. Heterodimers of CD51 with several integrin β chains function as receptors for extracellular matrix proteins. CD51/CD61 ( α V β 3 integrin, vitronectin receptor) mediates adhesion to fibronectin, fibrinogen, vitronectin, thrombospondin, von Willebrand factor, and CD31 (PECAM-1). CD51 is expressed on activated T lymphocytes, polymorphonuclear granulocytes, blastocysts, and osteoclasts. CD51 is reportedly not detectable on mouse platelets using either H9.2B8 or RMV-7 antibody clones. CD51 also forms heterodimers with CD29 (integrin β 1), integrins β 5, β

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD51
Applications Flow Cytometry
Classification Monoclonal
Clone RMV-7
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Itgav; Integrin alpha-V; Integrin αV ; VNRA; Vitronectin receptor alpha
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse (BALB/c) IL-2-activated killer (LAK) Cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.