Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat anti-Mouse IgG (Heavy chain) Secondary Antibody

Catalog No. MA180696
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA180696 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA180696 Supplier Invitrogen™ Supplier No. MA180696
Add to Cart
Add to Cart

Cocktail Secondary Antibody

Rat anti Mouse IgG Heavy Chain antibody cocktail has been shown to be specific for the gamma heavy chain of mouse immunoglobulin by immunoblotting techniques. It recognizes IgG2a and 2b strongly and IgG1 and IgG3 more weakly.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG (Heavy chain)
Applications ELISA
Classification Cocktail
Clone Cocktail
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Host Species Rat
Immunogen Purified IgG from BALB/c mice.
Purification Method Affinity chromatography
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype Ig
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.