Learn More
Rat anti-Mouse IgG1 Heavy Chain Secondary Antibody (LO-MG1-2), DyLight 550, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG1 Heavy Chain |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | LO-MG1-2 |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Host Species | Rat |
| Immunogen | Purified mouse IgG1 from BALB/c mice |
| Purification Method | Affinity purified |
| Quantity | 0.25 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.