Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-Mouse IgG1 Heavy Chain Secondary Antibody (LO-MG1-2), R&D Systems™
SDP

Catalog No. NB002835 Shop All Novus Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002835 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002835 Supplier R&D Systems Supplier No. NBP268427250UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rat Monoclonal Antibody

IgG1 Heavy Chain Monoclonal specifically detects IgG1 Heavy Chain in Mouse samples. It is validated for ELISA.

Specifications

Antigen IgG1 Heavy Chain
Applications ELISA
Classification Monoclonal
Clone LO-MG1-2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA
Formulation PBS
Host Species Rat
Immunogen Purified mouse IgG1 from BALB/c mice
Purification Method Affinity Purified
Quantity 250 μg
Regulatory Status RUO
Test Specificity Rat anti Mouse IgG1 Heavy Chain antibody, clone LO-MG1-2, recognizes murine IgG1 and does not bind other mouse immunoglobulin classes or subclasses.
Target Species Mouse
Content And Storage Store at 4°C.
Product Type Antibody
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.