Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-Mouse IgG2a Heavy Chain Secondary Antibody (LO-MG2a-7), R&D Systems™
SDP

Catalog No. NB002837 Shop All Novus Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002837 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002837 Supplier R&D Systems Supplier No. NBP268435250UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rat Monoclonal Antibody

IgG2a Heavy Chain Monoclonal specifically detects IgG2a Heavy Chain in Mouse samples. It is validated for ELISA.

Specifications

Antigen IgG2a Heavy Chain
Applications ELISA
Classification Monoclonal
Clone LO-MG2a-7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA
Formulation PBS
Host Species Rat
Immunogen Purified mouse IgG from BALB/c mice
Purification Method Affinity Purified
Quantity 250 μg
Regulatory Status RUO
Test Specificity Rat anti Mouse IgG2a-Biotin, clone LO-MG2a-7, recognizes the gamma 2a heavy chain of mouse immunoglobulin and does not cross-react with other murine immunoglobulin classes or subclasses. The antibody recognizes an allotypic determinant upon mouse IgG2a. It recognizes the IgHIa allotype (as expressed in Balb/c mice), but not the IgHIb allotype (as expressed in C57/BL mice).
Target Species Mouse
Content And Storage Store at 4°C.
Product Type Antibody
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.