Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Anti-Mouse IgG2A PE-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. F0129 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0129 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. F0129 Supplier R&D Systems Supplier No. F0129
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

IgG2a Monoclonal specifically detects IgG2a in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IgG2a
Applications Flow Cytometry
Classification Monoclonal
Clone 344701
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Host Species Rat
Immunogen Cocktail of mouse IgG 1 and IgG 2A
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity The specificity of this antibody was confirmed by direct ELISA on mouse IgG 2A monoclonal antibodies, and it does not cross-react with IgG 1 IgG 2B IgG 3 IgM, IgA, or IgE antibodies. By flow cytometry, this isotype-specific secondary antibody detects primary antibodies of the mouse IgG 2A isotype, and does not detect primary antibodies of mouse IgG 1 IgG 2B or IgG 3 class.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.