Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-Mouse IgG2b Heavy Chain Secondary Antibody (LO-MG2b-2), DyLight 650, R&D Systems™
SDP

Catalog No. NB554808 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB554808 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB554808 Supplier R&D Systems Supplier No. NBP268438C

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rat Monoclonal Antibody

IgG2b Heavy Chain Monoclonal antibody specifically detects IgG2b Heavy Chain in Mouse samples. It is validated for ELISA

Specifications

Antigen IgG2b Heavy Chain
Applications ELISA
Classification Monoclonal
Clone LO-MG2b-2
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Host Species Rat
Immunogen Purified mouse IgG2b
Purification Method Protein A purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.