Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-Mouse IgM Heavy Chain Secondary Antibody (eB121-15F9), Janelia Fluor 549, Novus Biologicals™
SDP

Catalog No. NB537636 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB537636 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB537636 Supplier Novus Biologicals Supplier No. NBP143282JF549
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IgM Heavy Chain Monoclonal antibody specifically detects IgM Heavy Chain in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen IgM Heavy Chain
Applications Flow Cytometry
Classification Monoclonal
Clone eB121-15F9
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain
Host Species Rat
Immunogen Mouse IgM
Purification Method Protein A or G purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3507
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.