Learn More
AbD Serotec RAT ANTI MOUSE NEUTROPHILS MAB
Supplier:  AbD Serotec MCA771GA
Encompass
Rat Anti Mouse Neutrophils, Monolconal, Clone: 7/4, 0.1mg, Purified IgG - liquid, Isotype: IgG 2a (Rat), Store at +4 deg.C, Application: FC, ELISA, IP, WB
Catalog No. NC9145221
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.