Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A9 Rat anti-Mouse, Alexa Fluor 647, Clone: 2B10, BD

Catalog No. BDB565833 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565833 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565833 Supplier BD Biosciences Supplier No. 565833
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 2B10 monoclonal antibody specifically binds to S100 calcium-binding protein A9 (S100A9), which is also known as Migration inhibitory factor-related protein 14 (MRP-14) or Calgranulin-B (CAGB). S100A9 is a member of the S100 family of calcium and zinc binding proteins. S100A9 is predominantly expressed in myeloid cells, such as, neutrophils and monocytes. It is one of the most abundant proteins expressed in myeloid cells. Similar to other S100 family of proteins, S100A9 functions as a calcium sensor and may enhance the chemotaxis and proinflammatory response of monocytes and neutrophils.

TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A9
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 2B10
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description S100a9; MRP-14; Calgranulin-B; GAGB; Cagb; BEE22; L1Ag; 60B8Ag; AW546964
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse S100A9 Recombinant Protein
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.