Learn More
Sino Biological Rat APOH PAb 100UL 80701-T16-100
Supplier:  Sino Biological 80701T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Apolipoprotein H / APOH (rR Apolipoprotein H / APOH; Catalog#80701-R08H; NP_001009626.1; Met1-Cys345). Apolipoprotein H / APOH specific IgG was purified by Rat Apolipoprotein H / APOH affinity chromatography.
Catalog No. 50-240-7681
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.