Learn More
Biomatik Corporation Rat ATPase, H+ Transporting, Lysosomal Accessory Protein 2 (ATP6AP2) ELISA Kit
Supplier:  Biomatik Corporation EKU02629
Encompass_Preferred
Rat ATPase, H+ Transporting, Lysosomal Accessory Protein 2 (ATP6AP2) ELISA Kit for detecting ATP6AP2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6AXS4. Gene ID: 302526. Target Synonyms: ATP6IP2; CAPER; ELDF10; XMRE; Renin/prorenin receptor; Embryonic liver differentiation factor 10; Vacuolar ATP synthase membrane sector-associated protein M8-9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4918
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.