Learn More
Immunology Consultants Rat Beta 2-Microglobulin ELISA Kit
SDP
Supplier:  Immunology Consultants E25B2
Encompass_Preferred
The Rat Beta 2-Microglobulin (B2M) test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring B2M in rat biological samples. B2M is a component of the major histocompatibility complex involving the presentation of peptide antigens to the immune system.
Catalog No. 50-240-6578
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.