Learn More
Novus Biologicals™ Rat beta Galactosidase ELISA Kit (Chemiluminescence)
Shop All R&D Systems Products

Specifications
Specifications
| Assay Range | 31.25 to 2000 pg/mL |
| Assay Sensitivity | 18.75 pg/mL |
| Detection Method | Chemiluminescence |
| Sample Type | Serum, Plasma, Other Biological Fluids |
| Specificity | This kit recognizes Rat beta Galactosidase in samples. No significant cross-reactivity or interference between Rat beta Galactosidase and analogues was observed |
| Target Species | Rat |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Synonym | EC 3.2.1.23; b-gal |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.